Product Details

SNP ID
rs118117111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167157399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATGAGCAAGCCCGGGCAGGTGA[A/G]GGCGACCATCAGGAGCCAGACGAGG
Phenotype
MIM: 602043
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR31 PubMed Links

Gene Details

Gene
GPR31
Gene Name
G protein-coupled receptor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005299.2 433 Missense Mutation CTC,TTC L145F NP_005290.2
Gene
LOC105378123
Gene Name
uncharacterized LOC105378123
There are no transcripts associated with this gene.

View Full Product Details