Product Details

SNP ID
rs116651711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34712456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCTCAAGCCTCACAGAACTGC[A/C]ATCAAGTGCCAACTATAAATAATAC
Phenotype
MIM: 601443
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFL2 PubMed Links

Gene Details

Gene
CFL2
Gene Name
cofilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243645.1 989 UTR 3 NP_001230574.1
NM_021914.7 989 UTR 3 NP_068733.1
NM_138638.4 989 UTR 3 NP_619579.1
XM_011536363.2 989 UTR 3 XP_011534665.1

View Full Product Details