Product Details

SNP ID
rs117497269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20472290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATGGGAGCAGAATTCACCTTG[A/T]TATTTTTTCTCCCCCAGATACACCT
Phenotype
MIM: 164050
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PNP PubMed Links

Gene Details

Gene
PNP
Gene Name
purine nucleoside phosphorylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000270.3 Intron NP_000261.2

View Full Product Details