Product Details

SNP ID
rs113644667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:68792646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCCTGCCAGGCAAACTTCGCC[C/T]CTCAAAACCCTGGCCTCCAGATTCA
Phenotype
MIM: 601064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP36L1 PubMed Links

Gene Details

Gene
ZFP36L1
Gene Name
ZFP36 ring finger protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244698.1 Intron NP_001231627.1
NM_001244701.1 Intron NP_001231630.1
NM_004926.3 Intron NP_004917.2

View Full Product Details