Product Details

SNP ID
rs112596178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70523286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTACAGGAGATCCCGTCCTGC[A/G]CATACACATCATCTGGGCATTGATG
Phenotype
MIM: 603712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM20 PubMed Links

Gene Details

Gene
ADAM20
Gene Name
ADAM metallopeptidase domain 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003814.4 10763 Missense Mutation GCG,GTG A541V NP_003805.3
XM_005268151.3 10763 Missense Mutation GCG,GTG A541V XP_005268208.1

View Full Product Details