Product Details

SNP ID
rs116002611
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70584212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCTGAAAAATATCAGTTATGA[C/T]GAAGAAAAAAGTCATGATGAAGCAA
Phenotype
MIM: 602984
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MED6 PubMed Links

Gene Details

Gene
MED6
Gene Name
mediator complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284209.1 1395 UTR 3 NP_001271138.1
NM_001284210.1 1395 UTR 3 NP_001271139.1
NM_001284211.1 1395 Intron NP_001271140.1
NM_005466.3 1395 UTR 3 NP_005457.2

View Full Product Details