Product Details

SNP ID
rs117112831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102376743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATGGCATCGATATCAGAGCCT[C/G]TTACATTCAGAGAGTTCTGCCCGTT
Phenotype
MIM: 615000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TECPR2 PubMed Links

Gene Details

Gene
TECPR2
Gene Name
tectonin beta-propeller repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172631.2 299 Missense Mutation CTT,GTT L8V NP_001166102.1
NM_014844.4 299 Missense Mutation CTT,GTT L8V NP_055659.2

View Full Product Details