Product Details

SNP ID
rs117017664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104582877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGCCTCCAGTTACACACTGTCC[C/T]TGCCAGAGGCCCCAGGCCTCCCCGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf180 PubMed Links

Gene Details

Gene
C14orf180
Gene Name
chromosome 14 open reading frame 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008404.2 Intron NP_001008404.1
NM_001286399.1 Intron NP_001273328.1
NM_001286400.1 Intron NP_001273329.1
XM_005267638.3 Intron XP_005267695.1
XM_011536764.2 Intron XP_011535066.1
Gene
TMEM179
Gene Name
transmembrane protein 179
There are no transcripts associated with this gene.

View Full Product Details