Product Details

SNP ID
rs112159330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54363018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTGCTTAGCTCTCTTGCTGTT[A/G]ATCTGGTCTCCCACTTCTGATGAGT
Phenotype
MIM: 606383 MIM: 609124
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR84 PubMed Links

Gene Details

Gene
GPR84
Gene Name
G protein-coupled receptor 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020370.2 1623 Silent Mutation ATC,ATT I278I NP_065103.1
XM_011538495.2 1623 Silent Mutation ATC,ATT I278I XP_011536797.1
Gene
LOC102724050
Gene Name
uncharacterized LOC102724050
There are no transcripts associated with this gene.

Gene
ZNF385A
Gene Name
zinc finger protein 385A
There are no transcripts associated with this gene.

View Full Product Details