Product Details

SNP ID
rs114420888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55452396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAACTCCTATGTACTTCTTTCT[C/T]AGAAATTTTTCCTTCTTAGAAGTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C2 PubMed Links

Gene Details

Gene
OR6C2
Gene Name
olfactory receptor family 6 subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054105.1 183 Silent Mutation CTC,CTT L61L NP_473446.1

View Full Product Details