Product Details

SNP ID
rs115027388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64193072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTGATAAACAGAAAGCGAAGC[A/G]GATAATATGCTGAAGAATGAAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf66 PubMed Links

Gene Details

Gene
C12orf66
Gene Name
chromosome 12 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300940.1 2008 Intron NP_001287869.1
NM_001300941.1 2008 UTR 3 NP_001287870.1
NM_152440.4 2008 UTR 3 NP_689653.3

View Full Product Details