Product Details

SNP ID
rs116977231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65293102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTTTTTTGCCTGAACTTAAGC[A/C]TCCCCACCTTGTTACCTAGCTTAAT
Phenotype
MIM: 613719
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MSRB3 PubMed Links

Gene Details

Gene
MSRB3
Gene Name
methionine sulfoxide reductase B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031679.2 Intron NP_001026849.1
NM_001193460.1 Intron NP_001180389.1
NM_001193461.1 Intron NP_001180390.1
NM_198080.3 Intron NP_932346.1

View Full Product Details