Product Details

SNP ID
rs117051519
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98455296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTGGTTAGCGGGGAGCCGCTG[A/T]GTGTGAGGATGGGCTTCAGGTAATT
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
SLC9A7P1 PubMed Links

Gene Details

Gene
SLC9A7P1
Gene Name
solute carrier family 9 member 7 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details