Product Details
- SNP ID
-
rs115298393
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108523542 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGGGGCGGCAACTGCAGGAGCC[A/G]CGGCAGGGCCGTTCTCAGCCTGAGG
- Phenotype
-
MIM: 611684
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FICD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35517322] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FICD
- Gene Name
- FIC domain containing
There are no transcripts associated with this gene.
- Gene
- SART3
- Gene Name
- squamous cell carcinoma antigen recognized by T-cells 3
View Full Product Details