Product Details
- SNP ID
-
rs116795662
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:107708047 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAAACACAAGGAGATTAATGGG[A/C]ATTCCTGTATAACAATGCTCGTTCT
- Phenotype
-
MIM: 602203
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLN
PubMed Links
Gene Details
- Gene
- SLN
- Gene Name
- sarcolipin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003063.2 |
|
Intron |
|
|
NP_003054.1 |
View Full Product Details