Product Details

SNP ID
rs113696572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAAACATCATCTCCTACGCAG[A/G]GTGCATGTCACAGCTCTACTTCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 338 Missense Mutation GAG,GGG E113G NP_001005194.1

View Full Product Details