Product Details

SNP ID
rs113972458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:133916652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGCCACAGCAGTGAAAGGAGAG[C/T]GGGCTTCTCTTGGATCTGGCACAGT
Phenotype
MIM: 613773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF9B PubMed Links

Gene Details

Gene
IGSF9B
Gene Name
immunoglobulin superfamily member 9B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277285.1 Intron NP_001264214.1
XM_006718795.3 Intron XP_006718858.1
XM_011542691.2 Intron XP_011540993.1
XM_011542692.2 Intron XP_011540994.1
XM_011542694.2 Intron XP_011540996.1
XM_011542695.2 Intron XP_011540997.1
XM_017017393.1 Intron XP_016872882.1
XM_017017394.1 Intron XP_016872883.1
XM_017017395.1 Intron XP_016872884.1
XM_017017396.1 Intron XP_016872885.1

View Full Product Details