Product Details

SNP ID
rs114031671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115579893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGACGTCTGAACACCACTCCA[C/T]GAAGTCCCTAACTACCACACTATCA
Phenotype
MIM: 608321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TICAM2 PubMed Links

Gene Details

Gene
TICAM2
Gene Name
toll like receptor adaptor molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021649.7 1806 UTR 3 NP_067681.1
Gene
TMED7-TICAM2
Gene Name
TMED7-TICAM2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164468.3 1806 UTR 3 NP_001157940.1
NM_001164469.3 1806 UTR 3 NP_001157941.1

View Full Product Details