Product Details

SNP ID
rs114025983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134404784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTTCTAACTTTTCTTTCAAA[C/T]TTCTTTTTCTTTCTTTTTTTTGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2AIPNL PubMed Links

Gene Details

Gene
CDKN2AIPNL
Gene Name
CDKN2A interacting protein N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080656.2 Intron NP_542387.1
XM_011543709.2 Intron XP_011542011.1
Gene
LOC105379186
Gene Name
uncharacterized LOC105379186
There are no transcripts associated with this gene.

View Full Product Details