Product Details

SNP ID
rs115361560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:358064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAACTTTCAAATGACTTACTTTT[A/C]AGTTTGCTTATTTTTTTTTCCTGTA
Phenotype
MIM: 194648
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR571 PubMed Links

Gene Details

Gene
MIR571
Gene Name
microRNA 571
There are no transcripts associated with this gene.

Gene
ZNF141
Gene Name
zinc finger protein 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003441.2 Intron NP_003432.1
XM_011513562.2 Intron XP_011511864.1
XM_011513563.2 Intron XP_011511865.1
XM_017008591.1 Intron XP_016864080.1

View Full Product Details