Product Details

SNP ID
rs111264107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71405044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAAAGGGCAGCCGCGACTCT[A/G]TCTGGGTTTCCATTTGTATCACTAA
Phenotype
MIM: 605516
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH23 PubMed Links

Gene Details

Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 Intron NP_001165401.1
NM_001171931.1 Intron NP_001165402.1
NM_001171932.1 Intron NP_001165403.1
NM_001171933.1 Intron NP_001165404.1
NM_001171934.1 Intron NP_001165405.1
NM_001171935.1 Intron NP_001165406.1
NM_001171936.1 Intron NP_001165407.1
NM_022124.5 Intron NP_071407.4
NM_052836.3 Intron NP_443068.1
XM_006717940.3 Intron XP_006718003.1
XM_006717942.3 Intron XP_006718005.1
XM_011540039.2 Intron XP_011538341.1
XM_011540042.2 Intron XP_011538344.1
XM_011540043.2 Intron XP_011538345.1
XM_011540044.2 Intron XP_011538346.1
XM_011540045.2 Intron XP_011538347.1
XM_011540046.2 Intron XP_011538348.1
XM_011540047.2 Intron XP_011538349.1
XM_011540048.2 Intron XP_011538350.1
XM_011540049.2 Intron XP_011538351.1
XM_011540051.2 Intron XP_011538353.1
XM_011540052.2 Intron XP_011538354.1
XM_017016499.1 Intron XP_016871988.1
XM_017016500.1 Intron XP_016871989.1
XM_017016501.1 Intron XP_016871990.1
XM_017016502.1 Intron XP_016871991.1
XM_017016503.1 Intron XP_016871992.1
XM_017016504.1 Intron XP_016871993.1
XM_017016505.1 Intron XP_016871994.1
XM_017016506.1 Intron XP_016871995.1
XM_017016507.1 Intron XP_016871996.1
XM_017016508.1 Intron XP_016871997.1

View Full Product Details