Product Details

SNP ID
rs112090972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102744072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGATGGAGAGGAGAAGGCTC[C/T]TGGGTGGCATGGCGCTCCTGCTCCT
Phenotype
MIM: 615570 MIM: 611129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFXN2 PubMed Links

Gene Details

Gene
SFXN2
Gene Name
sideroflexin 2
There are no transcripts associated with this gene.

Gene
WBP1L
Gene Name
WW domain binding protein 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083913.1 103 Silent Mutation CTG,TTG L7L NP_001077382.1
NM_017787.4 103 Intron NP_060257.4
XM_011539913.2 103 Intron XP_011538215.1
XM_017016360.1 103 UTR 5 XP_016871849.1

View Full Product Details