Product Details

SNP ID
rs113194493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:61904178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGATCTGGAAAAGGGAAGGAT[C/T]AAATATCCAGAAAAGAACTGGTGCT
Phenotype
MIM: 608538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID5B PubMed Links

Gene Details

Gene
ARID5B
Gene Name
AT-rich interaction domain 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244638.1 Intron NP_001231567.1
NM_032199.2 Intron NP_115575.1
XM_011540262.1 Intron XP_011538564.1
XM_017016765.1 Intron XP_016872254.1

View Full Product Details