Product Details

SNP ID
rs114393346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95606706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAGCGGGCTCTCTCCTGAGAT[A/T]AGACAAGTTTAACGTGAAGACCTTT
Phenotype
MIM: 138250
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH18A1 PubMed Links

Gene Details

Gene
ALDH18A1
Gene Name
aldehyde dehydrogenase 18 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017423.1 2706 UTR 3 NP_001017423.1
NM_001323412.1 2706 UTR 3 NP_001310341.1
NM_001323413.1 2706 UTR 3 NP_001310342.1
NM_001323414.1 2706 UTR 3 NP_001310343.1
NM_001323415.1 2706 UTR 3 NP_001310344.1
NM_001323416.1 2706 UTR 3 NP_001310345.1
NM_001323417.1 2706 UTR 3 NP_001310346.1
NM_001323418.1 2706 UTR 3 NP_001310347.1
NM_001323419.1 2706 UTR 3 NP_001310348.1
NM_002860.3 2706 UTR 3 NP_002851.2

View Full Product Details