Product Details

SNP ID
rs115228120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91809952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTGGGTCTTTGTGAGGGGAAG[A/G]AGGAGGATTCAACCAGAATAGTTTT
Phenotype
MIM: 607128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNKS2 PubMed Links

Gene Details

Gene
TNKS2
Gene Name
tankyrase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025235.3 Intron NP_079511.1
XM_005270185.4 Intron XP_005270242.2
XM_011540213.1 Intron XP_011538515.1
XM_011540215.2 Intron XP_011538517.1
XM_017016696.1 Intron XP_016872185.1
XM_017016697.1 Intron XP_016872186.1
XM_017016698.1 Intron XP_016872187.1
XM_017016699.1 Intron XP_016872188.1
XM_017016700.1 Intron XP_016872189.1
XM_017016701.1 Intron XP_016872190.1

View Full Product Details