Product Details

SNP ID
rs115706666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:90745130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTTATTACAGAAAGGTTGCCA[C/T]GTGCATATGTCTTAGCCTAGCCAGG
Phenotype
MIM: 182137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR7 PubMed Links

Gene Details

Gene
HTR7
Gene Name
5-hydroxytryptamine receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000872.4 Intron NP_000863.1
NM_019859.3 Intron NP_062873.1
NM_019860.3 Intron NP_062874.1

View Full Product Details