Product Details

SNP ID
rs116384695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119719654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTCCTCATGAGGGACTTTCCT[C/T]TTTCTTGGTCTGTGGTGCTTCTCAA
Phenotype
MIM: 609389
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5F PubMed Links

Gene Details

Gene
INPP5F
Gene Name
inositol polyphosphate-5-phosphatase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243194.1 Intron NP_001230123.1
NM_001243195.1 Intron NP_001230124.1
NM_014937.3 Intron NP_055752.1
XM_006717720.3 Intron XP_006717783.1
XM_011539524.2 Intron XP_011537826.1
XM_011539525.2 Intron XP_011537827.1
XM_011539527.2 Intron XP_011537829.2
XM_011539528.2 Intron XP_011537830.1
XM_011539529.1 Intron XP_011537831.1
XM_017015935.1 Intron XP_016871424.1
XM_017015936.1 Intron XP_016871425.1

View Full Product Details