Product Details

SNP ID
rs116903743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43849883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTGGTGCCACCTGCATATCT[A/G]AGCCCATGGAGTTCCTCTGGTCTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00619 PubMed Links

Gene Details

Gene
LINC00619
Gene Name
long intergenic non-protein coding RNA 619
There are no transcripts associated with this gene.

Gene
LINC00840
Gene Name
long intergenic non-protein coding RNA 840
There are no transcripts associated with this gene.

View Full Product Details