Product Details

SNP ID
rs138060113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110568595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCTTGGAGTCAGGAGAGCTGG[A/G]GCCCGTCGGCCTGCTTTTATAGACT
Phenotype
MIM: 606062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMC3 PubMed Links

Gene Details

Gene
SMC3
Gene Name
structural maintenance of chromosomes 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005445.3 Intron NP_005436.1

View Full Product Details