Product Details

SNP ID
rs138122775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133309407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCAGGATGGGGACGTGCAGCG[A/C]CACCTCTACCTCCAGGACGTGATCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TUBGCP2 PubMed Links

Gene Details

Gene
TUBGCP2
Gene Name
tubulin gamma complex associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256617.1 226 Intron NP_001243546.1
NM_001256618.1 226 Intron NP_001243547.1
NM_006659.3 226 Intron NP_006650.1
XM_017015595.1 226 Intron XP_016871084.1
XM_017015596.1 226 Intron XP_016871085.1
Gene
ZNF511
Gene Name
zinc finger protein 511
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145806.3 226 Silent Mutation CGA,CGC R57R NP_665805.2

View Full Product Details