Product Details

SNP ID
rs138700544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:90743651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCACACAGATACCGGTGGCCT[C/G]TTTTCTGGTCTCAACAGCACCCTCC
Phenotype
MIM: 182137
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HTR7 PubMed Links

Gene Details

Gene
HTR7
Gene Name
5-hydroxytryptamine receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000872.4 1578 Intron NP_000863.1
NM_019859.3 1578 Missense Mutation AAC,AAG N445K NP_062873.1
NM_019860.3 1578 Intron NP_062874.1

View Full Product Details