Product Details

SNP ID
rs140625528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:11463359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCACGTTTGACACCCGCACCTC[C/G]GCAGGACCTGGGACGGTAACTGCGA
Phenotype
MIM: 605405
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP6NL PubMed Links

Gene Details

Gene
USP6NL
Gene Name
USP6 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080491.3 4995 Silent Mutation GCC,GCG A540A NP_001073960.1
NM_014688.3 4995 Silent Mutation GCC,GCG A523A NP_055503.1
XM_006717542.3 4995 Silent Mutation GCC,GCG A551A XP_006717605.1
XM_011519762.2 4995 Silent Mutation GCC,GCG A551A XP_011518064.1
XM_011519763.2 4995 Silent Mutation GCC,GCG A367A XP_011518065.1
XM_017016971.1 4995 Silent Mutation GCC,GCG A541A XP_016872460.1
XM_017016972.1 4995 Silent Mutation GCC,GCG A367A XP_016872461.1
XM_017016973.1 4995 Silent Mutation GCC,GCG A546A XP_016872462.1

View Full Product Details