Product Details

SNP ID
rs141458099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:29289217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGCCTCTGGGGCAGGCACCAGG[A/G]ATCTGCCTTTTCAGTAAGACCTAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYZL1 PubMed Links

Gene Details

Gene
LYZL1
Gene Name
lysozyme like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032517.4 167 Intron NP_115906.3
XM_005252627.3 167 Missense Mutation AAT,GAT N34D XP_005252684.1
XM_017016791.1 167 Missense Mutation AAT,GAT N34D XP_016872280.1

View Full Product Details