Product Details

SNP ID
rs141850840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35607852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGGCCCCCGCCGCTGCCCCGA[C/T]CCCCGGGAGCCGGCCTCCGGGCAGA
Phenotype
MIM: 611922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJD4 PubMed Links

Gene Details

Gene
GJD4
Gene Name
gap junction protein delta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153368.2 497 Silent Mutation GAC,GAT D113D NP_699199.2

View Full Product Details