Product Details

SNP ID
rs142273937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102403397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGCCTGCCTTCAGCTTGACCC[A/G]AAGCAGCGCTGCATAGGTGCTGTAG
Phenotype
MIM: 164012 MIM: 602327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFKB2 PubMed Links

Gene Details

Gene
NFKB2
Gene Name
nuclear factor kappa B subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077494.3 3087 Intron NP_001070962.1
NM_001261403.2 3087 Intron NP_001248332.1
NM_001288724.1 3087 Intron NP_001275653.1
NM_001322934.1 3087 Intron NP_001309863.1
NM_001322935.1 3087 Intron NP_001309864.1
NM_002502.5 3087 Intron NP_002493.3
XM_011539830.2 3087 Intron XP_011538132.1
XM_011539831.2 3087 Intron XP_011538133.1
XM_017016278.1 3087 Intron XP_016871767.1
Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270965.1 3087 Missense Mutation CGG,TGG R960W NP_001257894.1
NM_001270966.1 3087 Missense Mutation CGG,TGG R581W NP_001257895.1
NM_002779.4 3087 Missense Mutation CGG,TGG R960W NP_002770.3
XM_011539968.1 3087 Missense Mutation CGG,TGG R960W XP_011538270.1
XM_011539969.2 3087 Missense Mutation CGG,TGG R960W XP_011538271.1
XM_017016433.1 3087 Missense Mutation CGG,TGG R960W XP_016871922.1
XM_017016434.1 3087 Missense Mutation CGG,TGG R960W XP_016871923.1

View Full Product Details