Product Details

SNP ID
rs143075956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94775154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTGGTGAAGGAAGCCCTGATT[C/G]ATCTTGGAGAGGAGTTTTCTGGAAG
Phenotype
MIM: 124020
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP2C19 PubMed Links

Gene Details

Gene
CYP2C19
Gene Name
cytochrome P450 family 2 subfamily C member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000769.2 290 Missense Mutation CAT,GAT H89D NP_000760.1

View Full Product Details