Product Details

SNP ID
rs143867323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101600803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGGCAGCTTGAAGTAGGAGAC[C/G]CAGCGCCCCTAGGAGCAGGGAACCT
Phenotype
MIM: 616467 MIM: 608071
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
FBXW4
Gene Name
F-box and WD repeat domain containing 4
There are no transcripts associated with this gene.

Gene
MIR3158-1
Gene Name
microRNA 3158-1
There are no transcripts associated with this gene.

Gene
MIR3158-2
Gene Name
microRNA 3158-2
There are no transcripts associated with this gene.

View Full Product Details