Product Details

SNP ID
rs145328984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94775106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCACTCTGTATTTTGGCCTGGAA[C/T]GCATGGTGGTGCTGCATGGATATGA
Phenotype
MIM: 124020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2C19 PubMed Links

Gene Details

Gene
CYP2C19
Gene Name
cytochrome P450 family 2 subfamily C member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000769.2 242 Missense Mutation CGC,TGC R73C NP_000760.1

View Full Product Details