Product Details

SNP ID
rs145346144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99784743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCACGTGTATAAATCCAGGACC[A/C]AGAGATCCTCTACCACCAAACTCTA
Phenotype
MIM: 601107
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCC2 PubMed Links

Gene Details

Gene
ABCC2
Gene Name
ATP binding cassette subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000392.4 393 Missense Mutation AAG,CAG K57Q NP_000383.1
XM_006717630.3 393 Intron XP_006717693.1
XM_006717631.3 393 Missense Mutation AAG,CAG K57Q XP_006717694.1
XM_011539291.2 393 Missense Mutation AAG,CAG K57Q XP_011537593.1
XM_017015675.1 393 Missense Mutation AAG,CAG K57Q XP_016871164.1

View Full Product Details