Product Details
- SNP ID
-
rs145680859
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:60871564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTAGTGCAATATCTTCCTTCTCT[C/T]GTTCCCTTTTCACACGCTGGTAGTG
- Phenotype
-
MIM: 607351
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RHOBTB1
PubMed Links
Gene Details
- Gene
- RHOBTB1
- Gene Name
- Rho related BTB domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242359.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
NP_001229288.1 |
NM_014836.4 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
NP_055651.1 |
XM_006718084.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
XP_006718147.1 |
XM_006718085.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
XP_006718148.1 |
XM_011540424.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
XP_011538726.1 |
XM_011540425.1 |
2318 |
UTR 3 |
|
|
XP_011538727.1 |
XM_011540426.1 |
2318 |
Intron |
|
|
XP_011538728.1 |
XM_011540428.1 |
2318 |
Intron |
|
|
XP_011538730.1 |
XM_017017000.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
XP_016872489.1 |
XM_017017001.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q670R |
XP_016872490.1 |
XM_017017002.1 |
2318 |
UTR 3 |
|
|
XP_016872491.1 |
XM_017017003.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872492.1 |
XM_017017004.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872493.1 |
XM_017017005.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872494.1 |
XM_017017006.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872495.1 |
XM_017017007.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872496.1 |
XM_017017008.1 |
2318 |
Missense Mutation |
CAA,CGA |
Q550R |
XP_016872497.1 |
View Full Product Details