Product Details

SNP ID
rs146739741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127137955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGTATGACCTGAGTTTCCTCA[A/G]TAGGGTGGAGTTTTCTTAGAACCGG
Phenotype
MIM: 601403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK1 PubMed Links

Gene Details

Gene
DOCK1
Gene Name
dedicator of cytokinesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290223.1 5449 Intron NP_001277152.1
NM_001380.4 5449 Intron NP_001371.1
XM_011539422.2 5449 Intron XP_011537724.1
XM_011539424.1 5449 Intron XP_011537726.1
XM_017015813.1 5449 Intron XP_016871302.1
XM_017015814.1 5449 Intron XP_016871303.1
XM_017015815.1 5449 Intron XP_016871304.1
XM_017015816.1 5449 Intron XP_016871305.1
XM_017015817.1 5449 Intron XP_016871306.1
Gene
FAM196A
Gene Name
family with sequence similarity 196 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039762.2 5449 Missense Mutation ACT,ATT T441I NP_001034851.1
XM_005252694.3 5449 Missense Mutation ACT,ATT T441I XP_005252751.1
XM_017016537.1 5449 Missense Mutation ACT,ATT T441I XP_016872026.1
XM_017016538.1 5449 Missense Mutation ACT,ATT T441I XP_016872027.1
XM_017016539.1 5449 Missense Mutation ACT,ATT T441I XP_016872028.1
XM_017016540.1 5449 Missense Mutation ACT,ATT T441I XP_016872029.1
XM_017016541.1 5449 Missense Mutation ACT,ATT T441I XP_016872030.1
XM_017016542.1 5449 Missense Mutation ACT,ATT T441I XP_016872031.1
XM_017016543.1 5449 Intron XP_016872032.1

View Full Product Details