Product Details

SNP ID
rs147673874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48434998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATACAGACAGCAGTCTAGAAG[C/T]AGCAGCTGGGCCTCTGGGCTGCTGT
Phenotype
MIM: 610585 MIM: 601158
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP22 PubMed Links

Gene Details

Gene
ARHGAP22
Gene Name
Rho GTPase activating protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256024.1 1485 Intron NP_001242953.1
NM_001256025.2 1485 Intron NP_001242954.1
NM_001256026.1 1485 Intron NP_001242955.1
NM_021226.3 1485 Intron NP_067049.2
XM_005270014.3 1485 Intron XP_005270071.1
XM_011540002.1 1485 Intron XP_011538304.1
XM_011540003.1 1485 Intron XP_011538305.1
XM_011540004.1 1485 Intron XP_011538306.1
XM_011540005.1 1485 Intron XP_011538307.1
XM_011540006.1 1485 Intron XP_011538308.1
XM_011540007.2 1485 Intron XP_011538309.1
XM_011540011.2 1485 Intron XP_011538313.1
XM_011540012.2 1485 Intron XP_011538314.1
XM_011540013.2 1485 Intron XP_011538315.1
XM_011540015.2 1485 Intron XP_011538317.1
XM_017016470.1 1485 Intron XP_016871959.1
XM_017016471.1 1485 Intron XP_016871960.1
XM_017016472.1 1485 Intron XP_016871961.1
XM_017016473.1 1485 Intron XP_016871962.1
XM_017016474.1 1485 Intron XP_016871963.1
XM_017016475.1 1485 Intron XP_016871964.1
XM_017016476.1 1485 Intron XP_016871965.1
XM_017016477.1 1485 Intron XP_016871966.1
Gene
MAPK8
Gene Name
mitogen-activated protein kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278547.1 1485 Missense Mutation GCA,GTA A418V NP_001265476.1
NM_001278548.1 1485 UTR 3 NP_001265477.1
NM_001323302.1 1485 UTR 3 NP_001310231.1
NM_001323320.1 1485 Missense Mutation GCA,GTA A342V NP_001310249.1
NM_001323321.1 1485 UTR 3 NP_001310250.1
NM_001323322.1 1485 Missense Mutation GCA,GTA A418V NP_001310251.1
NM_001323323.1 1485 Missense Mutation GCA,GTA A418V NP_001310252.1
NM_001323324.1 1485 UTR 3 NP_001310253.1
NM_001323325.1 1485 UTR 3 NP_001310254.1
NM_001323326.1 1485 UTR 3 NP_001310255.1
NM_001323327.1 1485 UTR 3 NP_001310256.1
NM_001323328.1 1485 Missense Mutation GCA,GTA A418V NP_001310257.1
NM_001323329.1 1485 Missense Mutation GCA,GTA A418V NP_001310258.1
NM_001323330.1 1485 Missense Mutation GCA,GTA A418V NP_001310259.1
NM_001323331.1 1485 Missense Mutation GCA,GTA A418V NP_001310260.1
NM_139046.3 1485 UTR 3 NP_620634.1
NM_139049.3 1485 Missense Mutation GCA,GTA A418V NP_620637.1

View Full Product Details