Product Details
- SNP ID
-
rs147722437
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:95237997 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGCTCCCGGGCATGCTTCTCACA[A/G]TAGATTTGATCCTCCACAAAGAAAT
- Phenotype
-
MIM: 605900
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACSM6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1049814] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACSM6
- Gene Name
- acyl-CoA synthetase medium-chain family member 6
There are no transcripts associated with this gene.
- Gene
- PDLIM1
- Gene Name
- PDZ and LIM domain 1
View Full Product Details