Product Details

SNP ID
rs147837408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69572934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCCCGTGTGCGAGTGGGGCTG[C/G]GCGGGGCGGACGTGGGGCAGGTCAC
Phenotype
MIM: 604882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NEUROG3 PubMed Links

Gene Details

Gene
NEUROG3
Gene Name
neurogenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020999.3 393 Missense Mutation CCC,CGC P37R NP_066279.2
XM_017016280.1 393 Missense Mutation CCC,CGC P37R XP_016871769.1

View Full Product Details