Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_005252384.2 | 2770 | UTR 3 | XP_005252441.1 | ||
XM_005252385.2 | 2770 | UTR 3 | XP_005252442.1 | ||
XM_005252386.2 | 2770 | UTR 3 | XP_005252443.1 | ||
XM_011519352.2 | 2770 | UTR 3 | XP_011517654.1 | ||
XM_011519353.2 | 2770 | UTR 3 | XP_011517655.1 | ||
XM_011519354.2 | 2770 | UTR 3 | XP_011517656.1 | ||
XM_011519355.2 | 2770 | UTR 3 | XP_011517657.1 | ||
XM_011519356.2 | 2770 | UTR 3 | XP_011517658.1 | ||
XM_017015837.1 | 2770 | UTR 3 | XP_016871326.1 | ||
XM_017015838.1 | 2770 | UTR 3 | XP_016871327.1 |