Product Details

SNP ID
rs148807437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119117606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGATATGCCAGAGTGAAGAAAA[C/T]GGCTCTTTCCTTAGTAAGGATTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM45A PubMed Links

Gene Details

Gene
FAM45A
Gene Name
family with sequence similarity 45 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303111.1 Intron NP_001290040.1
NM_001303112.1 Intron NP_001290041.1
NM_001303113.1 Intron NP_001290042.1
NM_207009.3 Intron NP_996892.1
XM_006717849.3 Intron XP_006717912.1
XM_006717850.3 Intron XP_006717913.1
XM_006717851.3 Intron XP_006717914.1
XM_006717852.3 Intron XP_006717915.1
XM_017016261.1 Intron XP_016871750.1
XM_017016262.1 Intron XP_016871751.1
XM_017016263.1 Intron XP_016871752.1
XM_017016264.1 Intron XP_016871753.1
XM_017016265.1 Intron XP_016871754.1
XM_017016266.1 Intron XP_016871755.1
XM_017016267.1 Intron XP_016871756.1

View Full Product Details