Product Details

SNP ID
rs148960265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27398645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCATAACCCTGGGACCATAATC[G/T]GAAAAATAATTCTCCTCTACGTTAA
Phenotype
MIM: 611791
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTCHD3 PubMed Links

Gene Details

Gene
PTCHD3
Gene Name
patched domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034842.3 2071 Silent Mutation TCA,TCC S651S NP_001030014.2

View Full Product Details