Product Details
- SNP ID
-
rs149237884
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:133356683 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGATGGACTCGTACTCCGTCCAC[A/G]CTGGGGTCTGCAGGCCCCTCTCCTT
- Phenotype
-
MIM: 602292
MIM: 609776
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ECHS1
PubMed Links
Gene Details
- Gene
- ECHS1
- Gene Name
- enoyl-CoA hydratase, short chain 1
There are no transcripts associated with this gene.
- Gene
- FUOM
- Gene Name
- fucose mutarotase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098483.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
NP_001091953.1 |
NM_001301827.1 |
305 |
Missense Mutation |
GCG,GTG |
A71V |
NP_001288756.1 |
NM_001301828.1 |
305 |
Missense Mutation |
GCG,GTG |
A49V |
NP_001288757.1 |
NM_198472.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
NP_940874.2 |
XM_011539670.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537972.1 |
XM_011539671.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537973.1 |
XM_011539672.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537974.1 |
XM_011539673.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537975.1 |
XM_011539674.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537976.1 |
XM_011539675.2 |
305 |
Missense Mutation |
GCG,GTG |
A94V |
XP_011537977.1 |
XM_017016094.1 |
305 |
Missense Mutation |
GCG,GTG |
A49V |
XP_016871583.1 |
- Gene
- PRAP1
- Gene Name
- proline rich acidic protein 1
There are no transcripts associated with this gene.
View Full Product Details