Product Details

SNP ID
rs149564225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7162714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAAAAGACTCACTCCCTGATCC[A/G]GCTACGATTAAACAACAACACTGGA
Phenotype
MIM: 615392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFMBT2 PubMed Links

Gene Details

Gene
SFMBT2
Gene Name
Scm-like with four mbt domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018039.1 3934 UTR 3 NP_001018049.1
NM_001029880.2 3934 UTR 3 NP_001025051.1
XM_005252551.2 3934 UTR 3 XP_005252608.1
XM_006717490.2 3934 UTR 3 XP_006717553.1
XM_011519611.1 3934 UTR 3 XP_011517913.1
XM_017016466.1 3934 UTR 3 XP_016871955.1

View Full Product Details