Product Details
- SNP ID
-
rs150145952
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29458536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCAGCGGGTAAATGGTTTTACA[C/G]AGCTTGGCTAAGACGTCTTCCACGA
- Phenotype
-
MIM: 604126
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SVIL
PubMed Links
Gene Details
- Gene
- SVIL
- Gene Name
- supervillin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001323599.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1842L |
NP_001310528.1 |
NM_001323600.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1758L |
NP_001310529.1 |
NM_003174.3 |
7209 |
Silent Mutation |
CTC,CTG |
L1726L |
NP_003165.2 |
NM_021738.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2152L |
NP_068506.2 |
XM_005252570.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2194L |
XP_005252627.1 |
XM_005252571.3 |
7209 |
Silent Mutation |
CTC,CTG |
L2194L |
XP_005252628.1 |
XM_005252573.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2194L |
XP_005252630.1 |
XM_011519630.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2253L |
XP_011517932.2 |
XM_011519631.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2247L |
XP_011517933.2 |
XM_011519632.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2222L |
XP_011517934.2 |
XM_011519633.2 |
7209 |
Silent Mutation |
CTC,CTG |
L2205L |
XP_011517935.2 |
XM_011519634.1 |
7209 |
Silent Mutation |
CTC,CTG |
L2194L |
XP_011517936.1 |
XM_011519635.1 |
7209 |
Silent Mutation |
CTC,CTG |
L2194L |
XP_011517937.1 |
XM_011519636.2 |
7209 |
Silent Mutation |
CTC,CTG |
L1969L |
XP_011517938.2 |
XM_011519637.2 |
7209 |
Silent Mutation |
CTC,CTG |
L1885L |
XP_011517939.2 |
XM_011519638.2 |
7209 |
Silent Mutation |
CTC,CTG |
L1853L |
XP_011517940.2 |
XM_017016575.1 |
7209 |
Silent Mutation |
CTC,CTG |
L2279L |
XP_016872064.1 |
XM_017016576.1 |
7209 |
Silent Mutation |
CTC,CTG |
L2252L |
XP_016872065.1 |
XM_017016577.1 |
7209 |
Silent Mutation |
CTC,CTG |
L2237L |
XP_016872066.1 |
XM_017016578.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1937L |
XP_016872067.1 |
XM_017016579.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1927L |
XP_016872068.1 |
XM_017016580.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1912L |
XP_016872069.1 |
XM_017016581.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1843L |
XP_016872070.1 |
XM_017016582.1 |
7209 |
Silent Mutation |
CTC,CTG |
L1811L |
XP_016872071.1 |
XM_017016583.1 |
7209 |
Intron |
|
|
XP_016872072.1 |
- Gene
- SVIL-AS1
- Gene Name
- SVIL antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details